4-(Trifluoromethoxy)aniline ≥98% for synthesis

Supplier: Matrix Scientific

005361-100G 005361-500G
101788-750EA 200 USD
101788-750 101788-752
4-(Trifluoromethoxy)aniline ≥98% for synthesis
4-(Trifluoromethoxy)aniline

Hazard code(s): Irritant

Formula: C₇H₆F₃NO
Boiling Pt: 73…75 °C (10mm)
Density: 1.31 g/cm³ (20 °C)
MDL Number: MFCD00041314
CAS Number: 461-82-5
EINECS: 207-317-5
UN: 2810
ADR: 6.1,II

Order Now

Specification Test Results

Boiling point 73-75°C/10m
Density 1.31
ND20 1.463


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR