3-(Trifluoromethoxy)phenol ≥97%

Supplier: Matrix Scientific

Synonyms: α,α,α-Trifluoro-3-hydroxyanisole

005358-5G 005358-25G
101788-742EA 26.15 USD
101788-742 101788-744
3-(Trifluoromethoxy)phenol ≥97%
3-(Trifluoromethoxy)phenol

Hazard code(s): Irritant

Formula: C₇H₅F₃O₂
MW: 178.11 g/mol
Boiling Pt: 69…70 °C (12mm)
Density: 1.37 g/cm³ (20 °C)
MDL Number: MFCD00040987
CAS Number: 827-99-6

Order Now

Specification Test Results

Boiling point 69-70°C/12m
Density 1.379
ND20 1.446

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR