2-(Trifluoromethoxy)aniline ≥97% for synthesis

Supplier: Matrix Scientific

005356-25G 005356-100G
101788-736EA 18.46 USD
101788-736 101788-738
2-(Trifluoromethoxy)aniline ≥97% for synthesis
2-(Trifluoromethoxy)aniline

Hazard code(s): Irritant

Formula: C₇H₆F₃NO
Boiling Pt: 61…63 °C (15mm)
Density: 1.301 g/cm³ (20 °C)
MDL Number: MFCD00035959
CAS Number: 1535-75-7
EINECS: 216-257-9
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 61-63°C/15m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR