4-(Trifluoromethoxy)phenyl isothiocyanate ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-(Trifluoromethoxy)phenylisothiocyanate

005353-1G 005353-5G
101788-728EA 33.85 USD
101788-728 101788-730
4-(Trifluoromethoxy)phenyl isothiocyanate ≥97% for synthesis
4-(Trifluoromethoxy)phenyl isothiocyanate

Hazard code(s): Irritant

Formula: C₈H₄F₃NOS
Boiling Pt: 50…52 °C (1mm)
Storage Temperature: Refrigerator
MDL Number: MFCD00042412
CAS Number: 64285-95-6

Order Now

Specification Test Results

Boiling point 50-52°C/1mm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR