4-(Trifluoromethoxy)anisole ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Methoxy-4-(trifluoromethoxy)benzene
005349-5G
005349-25G
005349-100G
101788-714EA
18.46
USD
101788-714
101788-716
101788-718
4-(Trifluoromethoxy)anisole ≥98%
4-(Trifluoromethoxy)anisole
Hazard code(s): Irritant
Formula:
C₈H₇F₃O₂ MW: 192.14 g/mol Boiling Pt: 164 °C Density: 1.272 g/cm³ (20 °C) |
MDL Number:
MFCD00216942 CAS Number: 710-18-9 UN: 1993 ADR: 3,III |
Specification Test Results
Boiling point | 164°C |
Density | 1.266 |
ND20 | 1.432 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...