4-(Trifluoromethoxy)benzyl alcohol ≥97% for synthesis

Supplier: Matrix Scientific

005345-5G 005345-25G
101788-704EA 21.54 USD
101788-704 101788-706
4-(Trifluoromethoxy)benzyl alcohol ≥97% for synthesis
4-(Trifluoromethoxy)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₇F₃O₂
Boiling Pt: 53 - 54 °C (0.5 -rr)
Density: 1.32 g/cm³ (20 °C)
MDL Number: MFCD00036029
CAS Number: 1736-74-9

Order Now

Specification Test Results

Boiling point 108°C/25mm
Density 1.326
ND20 1.449


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR