4-(Trifluoromethoxy)benzenesulfonamide

Supplier: Matrix Scientific

005338-1G 005338-5G
101788-686EA 21.54 USD
101788-686 101788-688
4-(Trifluoromethoxy)benzenesulfonamide
4-(Trifluoromethoxy)benzenesulphonamide
Formula: C₇H₆F₃NO₃S
MW: 241.19 g/mol
MDL Number: MFCD00798381
CAS Number: 1513-45-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR