(2-(Trifluoromethoxy)phenyl)acetic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-(2-(Trifluoromethoxy)phenyl)acetic acid
005329-5G
005329-25G
101788-674EA
30.77
USD
101788-674
101788-676
(2-(Trifluoromethoxy)phenyl)acetic acid ≥98%
(2-(Trifluoromethoxy)phenyl)acetic acid
Hazard code(s): Irritant
Formula:
F₃COC₆H₄CH₂COOH MW: 220.15 g/mol Melting Pt: 54…56 °C |
MDL Number:
MFCD00236325 CAS Number: 220239-67-8 |
Specification Test Results
Melting point | 54-56°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...