4-(Trifluoromethoxy)benzotrifluoride

Supplier: Matrix Scientific

Synonyms: 1-(Trifluoromethoxy)-4-(trifluoromethyl)benzene, 1-(Trifluormethoxy)-4-(trifluormethyl)benzene

005325-1G 005325-5G
101788-670EA 24.62 USD
101788-670 101788-672
4-(Trifluoromethoxy)benzotrifluoride
4-(Trifluoromethoxy)benzotrifluoride
Formula: C₈H₄F₆O
MW: 230.11 g/mol
MDL Number: MFCD00190125
CAS Number: 80258-33-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR