2-(Trifluoromethoxy)benzamide ≥97%
Supplier: Matrix Scientific
005313-1G
005313-5G
101788-662EA
26.15
USD
101788-662
101788-664
2-(Trifluoromethoxy)benzamide ≥97%
2-(Trifluoromethoxy)benzamide
Hazard code(s): Irritant
Formula:
C₈H₆F₃NO₂ MW: 205.14 g/mol Melting Pt: 153…155 °C |
MDL Number:
MFCD00042407 CAS Number: 127979-74-2 |
Specification Test Results
Melting point | 153-155°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...