(4-(Trifluoromethoxy)phenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-(Trifluoromethoxy)phenylacetic acid

005310-1G 005310-5G 005310-25G
101788-650EA 16.92 USD
101788-650 101788-652 101788-654
(4-(Trifluoromethoxy)phenyl)acetic acid ≥98%
(4-(Trifluoromethoxy)phenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₄(OCF₃))CH₂COOH
MW: 220.15 g/mol
MDL Number: MFCD00061239
CAS Number: 4315-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR