3-(Trifluoromethoxy)benzyl alcohol ≥97%

Supplier: Matrix Scientific

005308-1G 005308-5G
101788-642EA 27.69 USD
101788-642 101788-644
3-(Trifluoromethoxy)benzyl alcohol ≥97%
3-(Trifluoromethoxy)benzyl alcohol

Hazard code(s): Irritant

Formula: C₈H₇F₃O₂
MW: 192.14 g/mol
MDL Number: MFCD00061270
CAS Number: 50823-90-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR