4-(Trifluoromethoxy)benzoic acid ≥97%

Supplier: Matrix Scientific
Warning

005307-25G 005307-100G
101788-638EA 64.62 USD
101788-638 101788-640
4-(Trifluoromethoxy)benzoic acid ≥97%
4-(Trifluoromethoxy)benzoic acid

Hazard code(s): Irritant

Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272 °C (1013 hPa)
Melting Pt: 153.4 °C
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now

Specification Test Results

Melting point 150-153°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR