4-(Trifluoromethoxy)benzamide ≥97%

Supplier: Matrix Scientific

005305-1G 005305-5G
101788-632EA 21.54 USD
101788-632 101788-634
4-(Trifluoromethoxy)benzamide ≥97%
4-(Trifluoromethoxy)benzamide

Hazard code(s): Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 155…156 °C
MDL Number: MFCD00041384
CAS Number: 456-71-3

Order Now

Specification Test Results

Melting point 155-156°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR