3-(Trifluoromethoxy)benzamide ≥97%

Supplier: Matrix Scientific

005304-1G 005304-5G
101788-628EA 90.77 USD
101788-628 101788-630
3-(Trifluoromethoxy)benzamide ≥97%
3-(Trifluoromethoxy)benzamide

Hazard code(s): Irritant

Formula: C₈H₆F₃NO₂
Melting Pt: 115…116 °C
MDL Number: MFCD00041512
CAS Number: 658-91-3

Order Now

Specification Test Results

Melting point 115-116°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR