(±)-3,4-Difluoromandelic acid ≥97%

Supplier: Matrix Scientific

005301-1G 005301-5G
101788-624EA 29.23 USD
101788-624 101788-626
(±)-3,4-Difluoromandelic acid ≥97%
(±)-3,4-Difluoromandelic acid

Hazard code(s): Irritant

Formula: C₈H₆F₂O₃
Melting Pt: 92…94 °C
MDL Number: MFCD00061299
CAS Number: 132741-29-8

Order Now

Specification Test Results

Melting point 92-94°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR