(±)-2,4-Difluoromandelic acid ≥97%

Supplier: Matrix Scientific

005300-5G 005300-25G
101788-620EA 69.23 USD
101788-620 101788-622
(±)-2,4-Difluoromandelic acid ≥97%
(±)-2,4-Difluoromandelic acid

Hazard code(s): Irritant

Formula: C₈H₆F₂O₃
Melting Pt: 122…124 °C
MDL Number: MFCD00061296
CAS Number: 132741-30-1

Order Now

Specification Test Results

Melting point 122-124°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR