(±)-3,5-Difluoromandelic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
3,5-Difluoromandelic acid, (±)-3,5-Difluorophenylglycolic acid
005299-5G
005299-25G
101788-616EA
120
USD
101788-616
101788-618
(±)-3,5-Difluoromandelic acid ≥97%
(±)-3,5-Difluoromandelic acid
Hazard code(s): Irritant
Formula:
C₈H₆F₂O₃ MW: 188.13 g/mol Melting Pt: 135…137 °C |
MDL Number:
MFCD00061300 CAS Number: 132741-31-2 |
Specification Test Results
Melting point | 135-137°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...