1,3-Difluoro-4-iodobenzene ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2,4-Difluoroiodobenzene, 2,4-Difluoro-1-iodobenzene
005294-25G
005294-100G
101788-608EA
29.23
USD
101788-608
101788-610
1,3-Difluoro-4-iodobenzene ≥98%
1,3-Difluoro-4-iodobenzene
Hazard code(s): Irritant
Formula:
C₆H₃F₂I MW: 239.99 g/mol Boiling Pt: 175 - 176 °C (1013 hPa) Density: 2 g/cm³ (20 °C) |
MDL Number:
MFCD00040151 CAS Number: 2265-93-2 |
Specification Test Results
Boiling point | 57-59°C/15m |
Density | 2.006 |
ND20 | 1.557 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...