1,2-Difluoro-4-iodobenzene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3,4-Difluoroiodobenzene

005293-25G 005293-100G
101788-604EA 44.62 USD
101788-604 101788-606
1,2-Difluoro-4-iodobenzene ≥98%
1,2-Difluoro-4-iodobenzene

Hazard code(s): Irritant

Formula: C₆H₃F₂I
MW: 239.99 g/mol
Boiling Pt: 177 °C (10 -rr)
Density: 1.99 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00044630
CAS Number: 64248-58-4

Order Now

Specification Test Results

Boiling point 177-178°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR