1,3-Dichloro-2-fluorobenzene ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 2,6-Dichlorofluorobenzene

005282-25G 005282-100G
101788-588EA 16.92 USD
101788-588 101788-590
1,3-Dichloro-2-fluorobenzene ≥99%
1,3-Dichloro-2-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃Cl₂F
MW: 164.99 g/mol
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: 39 °C
MDL Number: MFCD00010307
CAS Number: 2268-05-5
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Boiling point 81-82°C/45m
Melting point 37-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR