1,2-Dichloro-4-fluorobenzene ≥99%

Supplier: Matrix Scientific
Warning

005279-25G 005279-100G
101788-574EA 55.38 USD
101788-574 101788-576
1,2-Dichloro-4-fluorobenzene ≥99%
1,2-Dichloro-4-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃Cl₂F
Boiling Pt: 168 °C (1013 hPa)
Melting Pt: -1 °C
Density: 1.4 g/cm³ (20 °C)
MDL Number: MFCD00018119
CAS Number: 1435-49-0
EINECS: 215-858-3

Order Now

Specification Test Results

Boiling point 86°C/35mm
Melting point -1°C
Density 1.403
ND20 1.524


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR