2,6-Difluorobenzyl alcohol ≥98%

Supplier: Matrix Scientific
Warning

005245-5G 005245-25G 005245-100G
101788-532EA 18.46 USD
101788-532 101788-534 101788-536
2,6-Difluorobenzyl alcohol ≥98%
2,6-Difluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆F₂O
Boiling Pt: 88 °C (14mm)
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00004603
CAS Number: 19064-18-7
EINECS: 242-792-2

Order Now

Specification Test Results

Boiling point 88°C/14mm
Density 1.3
ND20 1.495


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR