2,4-Difluorobenzyl alcohol ≥99%

Supplier: Matrix Scientific
Warning

005243-5G 005243-25G 005243-100G
101788-526EA 20 USD
101788-526 101788-528 101788-530
2,4-Difluorobenzyl alcohol ≥99%
2,4-Difluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆F₂O
Boiling Pt: 52 °C (1mm)
Density: 1.290 g/cm³
MDL Number: MFCD00009983
CAS Number: 56456-47-4
EINECS: 260-191-3

Order Now

Specification Test Results

Boiling point 52°C/1mm
Density 1.29
ND20 1.487


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR