α-Bromo-2,5-difluorotoluene ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide

005237-25G 005237-100G
101788-522EA 30.77 USD
101788-522 101788-524
α-Bromo-2,5-difluorotoluene ≥98%
α-Bromo-2,5-difluorotoluene

Hazard code(s): Corrosive, Lachrymator

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.609 g/cm³ (20 °C)
MDL Number: MFCD00009897
CAS Number: 85117-99-3
EINECS: 285-651-0
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Density 1.609
ND20 1.526

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR