3,4-Difluorobenzyl alcohol ≥99%

Supplier: Matrix Scientific
Warning

005234-5G 005234-25G
101788-518EA 13.85 USD
101788-518 101788-520
3,4-Difluorobenzyl alcohol ≥99%
3,4-Difluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆F₂O
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.262 g/cm³
MDL Number: MFCD00010628
CAS Number: 85118-05-4
EINECS: 285-657-3

Order Now

Specification Test Results

Density 1.262
ND20 1.49


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR