2,5-Difluorobenzyl alcohol ≥99%

Supplier: Matrix Scientific
Danger

Synonyms: (2,5-Difluorophenyl)methanol

005232-5G 005232-25G
101788-514EA 20 USD
101788-514 101788-516
2,5-Difluorobenzyl alcohol ≥99%
2,5-Difluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.29 g/cm³ (20 °C)
MDL Number: MFCD00010627
CAS Number: 75853-20-2
EINECS: 278-325-4

Order Now

Specification Test Results

Density 1.29
ND20 1.49

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR