2,3-Difluorobenzyl alcohol ≥95%

Supplier: Matrix Scientific
Danger

005231-5G 005231-25G
101788-512EA 44.62 USD
101788-512 102550-854
2,3-Difluorobenzyl alcohol ≥95%
2,3-Difluorobenzyl alcohol

Hazard code(s): Irritant

Formula: C₇H₆F₂O
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.28 g/cm³ (20 °C)
MDL Number: MFCD00010312
CAS Number: 75853-18-8

Order Now

Specification Test Results

Density 1.282
ND20 1.492


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR