3,4-Dichlorobenzoylacetonitrile ≥96%

Supplier: Matrix Scientific

Synonyms: 3-(3,4-Dichlorophenyl)-3-oxopropanenitrile

005225-1G 005225-5G 005225-25G
101788-500EA 27.69 USD
101788-500 101788-502 101788-504
3,4-Dichlorobenzoylacetonitrile ≥96%
3,4-Dichlorobenzoylacetonitrile

Hazard code(s): Irritant

Formula: C₉H₅Cl₂NO
MW: 214.05 g/mol
Melting Pt: 114…117 °C
MDL Number: MFCD00067919
CAS Number: 4640-68-0

Order Now

Specification Test Results

Melting point 114-117°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR