3,5-Dichloro-α,α,α-trifluorotoluene ≥97%

Supplier: Matrix Scientific

005222-1G 005222-5G
101788-496EA 30.77 USD
101788-496 101788-498
3,5-Dichloro-α,α,α-trifluorotoluene ≥97%
3,5-Dichloro-α,α,α-trifluorotoluene

Hazard code(s): Corrosive

Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 173…174 °C
Melting Pt: –13 °C
Density: 1.478 g/cm³
MDL Number: MFCD00203929
CAS Number: 54773-20-5
EINECS: 259-337-9
UN: 2234
ADR: 3,III

Order Now

Specification Test Results

Boiling point 173-174°C
Melting point -13°C
Density 1.478

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR