2,3-Dichloro-α,α,α-trifluorotoluene ≥98%

Supplier: Matrix Scientific

005218-1G 005218-5G
101788-492EA 21.54 USD
101788-492 101788-494
2,3-Dichloro-α,α,α-trifluorotoluene ≥98%
2,3-Dichloro-α,α,α-trifluorotoluene

Hazard code(s): Irritant

Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Melting Pt: 18…19 °C
Density: 1.52 g/cm³
MDL Number: MFCD00069115
CAS Number: 54773-19-2
EINECS: 259-336-3
UN: 2234
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR