2,3-Difluorobenzophenone ≥97%

Supplier: Matrix Scientific

005216-1G 005216-5G
101788-488EA 29.23 USD
101788-488 101788-490
2,3-Difluorobenzophenone ≥97%
2,3-Difluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Boiling Pt: 176…178 °C (16mm)
MDL Number: MFCD00061139
CAS Number: 208173-20-0

Order Now

Specification Test Results

Boiling point 176-178°C/1
ND20 1.5685

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR