2,5-Difluorobenzophenone ≥98%
Supplier: Matrix Scientific
|
Warning
|
005214-5G
005214-25G
101788-480EA
38.46
USD
101788-480
101788-482
2,5-Difluorobenzophenone ≥98%
2,5-Difluorobenzophenone
Hazard code(s): Irritant
Formula:
C₁₃H₈F₂O MW: 218.2 g/mol Boiling Pt: 120…123 °C (0.35mm) Density: 1.262 g/cm³ |
MDL Number:
MFCD00009899 CAS Number: 85068-36-6 EINECS: 285-298-2 |
Specification Test Results
Boiling point | 120-123°C/0 |
Density | 1.262 |
ND20 | 1.5693 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...