2,5-Dichlorobenzophenone ≥98%

Supplier: Matrix Scientific

Synonyms: (2,5-Dichlorophenyl)(phenyl)methanone

005212-5G 005212-25G
101788-472EA 36.92 USD
101788-472 101788-474
2,5-Dichlorobenzophenone ≥98%
2,5-Dichlorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₈Cl₂O
MW: 251.11 g/mol
Melting Pt: 87…88 °C
MDL Number: MFCD00079746
CAS Number: 16611-67-9

Order Now

Specification Test Results

Melting point 87-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR