3,5-Difluorobenzophenone ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: (3,5-Difluorophenyl)(phenyl)methanone

005211-1G 005211-5G
101788-468EA 21.54 USD
101788-468 101788-470
3,5-Difluorobenzophenone ≥97%
3,5-Difluorobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₈F₂O
MW: 218.2 g/mol
Melting Pt: 57…59 °C
MDL Number: MFCD00061140
CAS Number: 179113-89-4

Order Now

Specification Test Results

Melting point 57-59°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR