2,5-Difluorobenzonitrile ≥98%

Supplier: Matrix Scientific

005208-25G 005208-100G
101788-460EA 129.23 USD
101788-460 101788-462
2,5-Difluorobenzonitrile ≥98%
2,5-Difluorobenzonitrile

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₇H₃F₂N
MW: 139.1 g/mol
Boiling Pt: 87 - 92 °C (30 -rr)
Melting Pt: 29.5 - 30.5 °C
MDL Number: MFCD00001777
CAS Number: 64248-64-2
EINECS: 264-753-9
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 188°C
Melting point 33-35°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR