3,5-Difluorobenzonitrile ≥98%
Supplier: Matrix Scientific
005207-25G
005207-100G
101788-456EA
60
USD
101788-456
101788-458
3,5-Difluorobenzonitrile ≥98%
3,5-Difluorobenzonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₇H₃F₂N Boiling Pt: 159 - 160 °C (1013 hPa) Melting Pt: 84…86 °C |
MDL Number:
MFCD00010311 CAS Number: 64248-63-1 EINECS: 264-752-3 UN: 3439 ADR: 6.1,III |
Specification Test Results
Melting point | 84-86°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...