3,5-Dichlorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

005203-25G 005203-100G
101788-444EA 20 USD
101788-444 101788-446
3,5-Dichlorobenzoic acid ≥98%
3,5-Dichlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 309 °C (1013 hPa)
Melting Pt: 183 - 185 °C
Storage Temperature: Ambient
MDL Number: MFCD00002494
CAS Number: 51-36-5
EINECS: 200-092-4

Order Now

Specification Test Results

Melting point 186-187°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR