2,4-Difluorobenzoic acid ≥98%
Supplier: Matrix Scientific
|
Warning
|
005198-25G
005198-100G
101788-430EA
41.54
USD
101788-430
101788-432
2,4-Difluorobenzoic acid ≥98%
2,4-Difluorobenzoic acid
Hazard code(s): Irritant
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Boiling Pt: 241 °C (1013 hPa) Melting Pt: 185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011670 CAS Number: 1583-58-0 EINECS: 216-430-9 |
Specification Test Results
Melting point | 188-190°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...