2,3-Difluorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

005197-25G 005197-100G
101788-426EA 120 USD
101788-426 101788-428
2,3-Difluorobenzoic acid ≥98%
2,3-Difluorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄F₂O₂
MW: 158.1 g/mol
Melting Pt: 163…165 °C
MDL Number: MFCD00010267
CAS Number: 4519-39-5

Order Now

Specification Test Results

Melting point 163-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR