2,6-Dichlorobenzoic acid ≥98%

Supplier: Matrix Scientific
Warning

005195-25G 005195-100G 005195-500G
101788-418EA 23.08 USD
101788-418 101788-420 101788-422
2,6-Dichlorobenzoic acid ≥98%
2,6-Dichlorobenzoic acid

Hazard code(s): Irritant

Formula: C₇H₄Cl₂O₂
MW: 191.01 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 138 - 139 °C
Storage Temperature: Ambient
MDL Number: MFCD00002418
CAS Number: 50-30-6
EINECS: 200-025-9

Order Now

Specification Test Results

Melting point 143-145°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR