3,5-Difluorobenzhydrazide

Supplier: Matrix Scientific

Synonyms: 3,5-Difluorobenzohydrazide, 3,5-Difluorobenzoic hydrazide, 3,5-Difluorobenzene-1-carbohydrazide, 3,5-Difluorobenzoic acid hydrazide

005190-1G 005190-5G
101788-406EA 52.31 USD
101788-406 101788-408
3,5-Difluorobenzhydrazide
3,5-Difluorobenzhydrazide

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₆F₂N₂O
MW: 172.13 g/mol
Melting Pt: 168…170 °C
MDL Number: MFCD01310810
CAS Number: 244022-63-7

Order Now

Specification Test Results

Melting point 168-170°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR