3,4-Difluorobenzhydrazide

Supplier: Matrix Scientific

005189-1G 005189-5G
101788-402EA 72.31 USD
101788-402 101788-404
3,4-Difluorobenzhydrazide
3,4-Difluorobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆F₂N₂O
Melting Pt: 143…145 °C
MDL Number: MFCD00672992
CAS Number: 229957-07-7

Order Now

Specification Test Results

Melting point 143-145°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR