3,4-Difluorobenzamide ≥97%

Supplier: Matrix Scientific

005170-1G 005170-5G
101788-380EA 15.38 USD
101788-380 101788-382
3,4-Difluorobenzamide ≥97%
3,4-Difluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Melting Pt: 134 °C
MDL Number: MFCD00015549
CAS Number: 85118-04-3
EINECS: 285-656-8

Order Now

Specification Test Results

Melting point 134°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR