2,3-Difluorobenzamide ≥97%

Supplier: Matrix Scientific

005169-5G 005169-25G
101788-376EA 63.08 USD
101788-376 101788-378
2,3-Difluorobenzamide ≥97%
2,3-Difluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
MDL Number: MFCD00061137
CAS Number: 18355-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR