2,6-Difluorobenzamide ≥97%

Supplier: Matrix Scientific

005168-25G 005168-100G
101788-372EA 24.62 USD
101788-372 101788-374
2,6-Difluorobenzamide ≥97%
2,6-Difluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 145…148 °C
MDL Number: MFCD00007972
CAS Number: 18063-03-1
EINECS: 241-972-8

Order Now

Specification Test Results

Melting point 145-148°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR