2,5-Difluorobenzamide ≥98%

Supplier: Matrix Scientific

005167-1G 005167-5G 005167-25G
101788-366EA 21.54 USD
101788-366 101788-368 101788-370
2,5-Difluorobenzamide ≥98%
2,5-Difluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅F₂NO
Melting Pt: 114…116 °C
MDL Number: MFCD00015548
CAS Number: 85118-03-2
EINECS: 285-655-2

Order Now

Specification Test Results

Melting point 114-116°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR