2,4-Difluorobenzamide ≥95%

Supplier: Matrix Scientific

005166-5G 005166-25G
101788-362EA 52.31 USD
101788-362 101788-364
2,4-Difluorobenzamide ≥95%
2,4-Difluorobenzamide

Hazard code(s): Irritant

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
Melting Pt: 160…162 °C
MDL Number: MFCD00015547
CAS Number: 85118-02-1
EINECS: 285-654-7

Order Now

Specification Test Results

Melting point 160-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR