2,3-Difluorobenzaldehyde ≥97%

Supplier: Matrix Scientific
Warning

005162-5G 005162-100G
101788-346EA 601.54 USD
101788-346 101788-350
2,3-Difluorobenzaldehyde ≥97%
2,3-Difluorobenzaldehyde

Hazard code(s): Flammable, Air Sensitive

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 64…65 °C (17mm)
Density: 1.3 g/cm³ (20 °C)
MDL Number: MFCD00010292
CAS Number: 2646-91-5
UN: 1989
ADR: 3,III

Order Now

Specification Test Results

Boiling point 64-65°C/17m
Density 1.301
ND20 1.499

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR