3,4-Difluorobenzaldehyde ≥97%

Supplier: Matrix Scientific

005159-5G 005159-25G 005159-100G
101788-330EA 10.77 USD
101788-330 101788-332 101788-334
3,4-Difluorobenzaldehyde ≥97%
3,4-Difluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 53 °C (1013 hPa)
Density: 1.29 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010328
CAS Number: 34036-07-2
EINECS: 422-180-3

Order Now

Specification Test Results

Density 1.288
ND20 1.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR